Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106797.3 | NP_001100267.1 | 4 | 378 | ||
XM_006242391.2 | XP_006242453.1 | 4 | 442 | |||
XM_006242393.2 | XP_006242455.1 | 3 | 337 | |||
XM_008765707.1 | XP_008763929.1 | 3 | 351 | |||
GenBank mRNA | BC166908.1 | - | 4 | 379 | 6889335 | |
BC168739.1 | - | 4 | 379 | 6918617 |