Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFAND5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102420.2 | NP_001095890.1 | 7 | 1261 | ||
NM_001102421.2 | NP_001095891.1 | 6 | 1124 | |||
NM_001278243.1 | NP_001265172.1 | 7 | 840 | |||
NM_001278244.1 | NP_001265173.1 | 6 | 703 | |||
NM_001278245.1 | NP_001265174.1 | 7 | 837 | |||
NM_006007.3 | NP_005998.1 | 6 | 1121 | |||
GenBank mRNA | AF062346.1 | - | 6 | 844 | ||
AF062347.1 | - | 6 | 699 | |||
AK025782.1 | - | 7 | 724 | |||
AK290849.1 | - | 6 | 844 | |||
AK307376.1 | - | 4 | 545 | |||
BC011018.1 | - | 6 | 960 | 4331933 | ||
BC027707.2 | - | 7 | 1211 | 3945047 | ||
BC073131.1 | - | 6 | 625 | 6137219 | ||
BX649046.1 | - | 1 | 649 | |||
X75687.1 | - | 1 | 68 |