Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kank2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270413.1 | NP_001257342.1 | 10 | 2419 | ||
XM_006242589.2 | XP_006242651.1 | 11 | 2824 | |||
XM_006242590.2 | XP_006242652.1 | 10 | 2679 |