Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1235 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000447.1 | NP_001000447.1 | 1 | 425 |
Gene Symbol: Olr1231 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000813.1 | NP_001000813.1 | 1 | 434 |