Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF226 SIRNA Assays ›
Gene Symbol:
ZNF226
Gene Name:
zinc finger protein 226
Gene Aliases:
-
Chromosome Location:
Chr.19: 44165062 - 44177685 on Build GRCh38
UniGene ID:
Hs.145956
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s15309
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF226
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001032372.1
NP_001027544.1
5
347
NM_001032373.1
NP_001027545.1
5
311
NM_001032374.1
NP_001027546.1
5
480
NM_001146220.2
NP_001139692.1
6
371
NM_001319088.1
NP_001306017.1
5
514
NM_001319089.1
NP_001306018.1
6
371
NM_001319090.1
NP_001306019.1
5
391
NM_015919.3
NP_057003.2
5
311
NM_016444.2
NP_057528.2
6
534
XM_005259227.2
XP_005259284.1
6
645
XM_006723367.3
XP_006723430.1
7
541
XM_006723368.3
XP_006723431.1
7
427
XM_006723369.2
XP_006723432.1
6
484
XM_017027262.1
XP_016882751.1
6
493
XM_017027263.1
XP_016882752.1
5
315
XM_017027264.1
XP_016882753.1
5
472
XM_017027265.1
XP_016882754.1
5
379
GenBank mRNA
AF070666.1
-
5
306
AF228418.1
-
6
532
AK023091.1
-
5
307
AK314554.1
-
5
306
AK314724.1
-
6
330
BC017786.1
-
5
303
BC024197.2
-
5
347
3532273
BI457780.1
-
5
312
BX091242.1
-
5
473
BX537487.1
-
5
303
BX648775.1
-
5
475
CD700800.1
-
5
337
DA080156.1
-
5
475
DA178589.1
-
5
334
DA435005.1
-
6
317
DA558473.1
-
5
319
DB015503.1
-
5
500
DB174587.1
-
6
484
DR002748.1
-
5
353
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0