Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1248 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000452.1 | NP_001000452.1 | 1 | 762 |
Gene Symbol: Olr1249 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001020.1 | NP_001001020.1 | 1 | 762 |