Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF227 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289166.1 | NP_001276095.1 | 5 | 431 | ||
NM_001289167.1 | NP_001276096.1 | 5 | 387 | |||
NM_001289168.1 | NP_001276097.1 | 4 | 366 | |||
NM_001289169.1 | NP_001276098.1 | 4 | 369 | |||
NM_182490.2 | NP_872296.1 | 5 | 428 | |||
XM_005259232.2 | XP_005259289.1 | 5 | 559 | |||
XM_011527289.2 | XP_011525591.1 | 5 | 707 | |||
XM_017027266.1 | XP_016882755.1 | 5 | 562 | |||
XM_017027268.1 | XP_016882757.1 | 4 | 497 | |||
XM_017027269.1 | XP_016882758.1 | 4 | 467 | |||
XM_017027270.1 | XP_016882759.1 | 4 | 645 | |||
XM_017027271.1 | XP_016882760.1 | 4 | 643 | |||
XM_017027275.1 | XP_016882764.1 | 5 | 530 | |||
XM_017027276.1 | XP_016882765.1 | 5 | 526 | |||
XM_017027277.1 | XP_016882766.1 | 4 | 390 | |||
GenBank mRNA | AK092253.1 | - | 5 | 428 | ||
AK299266.1 | - | 3 | 319 | |||
AK316048.1 | - | 5 | 377 | |||
AY166786.1 | - | 2 | 78 | |||
BC047570.1 | - | 5 | 418 | 4838846 | ||
DA494064.1 | - | 5 | 429 | |||
DA929840.1 | - | 4 | 359 |