Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF230 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006300.3 | NP_006291.2 | 5 | 1391 | ||
GenBank mRNA | BC050340.1 | - | 5 | 1368 | 4821663 | |
BC094708.1 | - | 5 | 1391 | 30402948 |