Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl9l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106817.1 | NP_001100287.1 | 9 | 2941 | ||
XM_006242923.2 | XP_006242985.1 | 8 | 4656 | |||
XM_006242924.2 | XP_006242986.1 | 7 | 4599 | |||
XM_006242925.2 | XP_006242987.1 | 7 | 4554 | |||
XM_006242928.2 | XP_006242990.1 | 5 | 2279 |