Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35f2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106822.1 | NP_001100292.1 | 5 | 856 | ||
XM_006243069.2 | XP_006243131.1 | 6 | 1100 |