Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106828.1 | NP_001100298.1 | 22 | 2162 | ||
XM_006243191.1 | XP_006243253.1 | 22 | 2446 | |||
XM_006243192.2 | XP_006243254.1 | 21 | 3104 | |||
XM_006243193.2 | XP_006243255.1 | 21 | 1973 | |||
XM_008766231.1 | XP_008764453.1 | 21 | 3101 | |||
XM_008766232.1 | XP_008764454.1 | 21 | 3044 | |||
XM_008766233.1 | XP_008764455.1 | 21 | 3041 | |||
GenBank mRNA | FQ223826.1 | - | 19 | 2682 |