Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106828.1 | NP_001100298.1 | 6 | 676 | ||
XM_006243191.1 | XP_006243253.1 | 6 | 957 | |||
XM_006243192.2 | XP_006243254.1 | 5 | 1615 | |||
XM_006243193.2 | XP_006243255.1 | 5 | 484 | |||
XM_008766231.1 | XP_008764453.1 | 5 | 1615 | |||
XM_008766232.1 | XP_008764454.1 | 5 | 1615 | |||
XM_008766233.1 | XP_008764455.1 | 5 | 1615 | |||
GenBank mRNA | BC167025.1 | - | 4 | 211 | 7365328 | |
FQ223826.1 | - | 5 | 1339 |