Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gramd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006243197.2 | XP_006243259.1 | 8 | 862 | ||
XM_006243198.2 | XP_006243260.1 | 9 | 1223 | |||
XM_006243199.2 | XP_006243261.1 | 8 | 1271 | |||
XM_006243200.2 | XP_006243262.1 | 8 | 1611 | |||
XM_006243201.2 | XP_006243263.1 | 8 | 3072 | |||
XM_008766374.1 | XP_008764596.1 | 8 | 742 | |||
XM_217153.7 | XP_217153.5 | 8 | 661 |