Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC30A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013309.5 | NP_037441.2 | 6 | 1300 | ||
XM_011521997.2 | XP_011520299.1 | 6 | 1804 | |||
XM_017022560.1 | XP_016878049.1 | 5 | 1106 | |||
GenBank mRNA | AF025409.1 | - | 5 | 988 | ||
AK290874.1 | - | 5 | 1106 | |||
AY029482.1 | - | 2 | 265 | |||
BC026089.2 | - | 6 | 1300 | 4829839 |