Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aph1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047090.1 | NP_001040555.1 | 4, 5 | 607 | ||
GenBank mRNA | BC098711.1 | - | 4, 5 | 469 | ||
BC166592.1 | - | 4, 5 | 563 | 7929867 | ||
DQ672552.1 | - | 4, 5 | 607 |
Gene Symbol: LOC100911293 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766401.1 | XP_008764623.1 | 602 |
Gene Symbol: LOC691658 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750532.3 | XP_003750580.1 | 4, 5 | 636 | ||
XM_006243318.2 | XP_006243380.1 | 4, 5 | 844 |