Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aph1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047090.1 | NP_001040555.1 | 6 | 761 | ||
GenBank mRNA | BC098711.1 | - | 6 | 623 | ||
BC166592.1 | - | 6 | 717 | 7929867 | ||
DQ672552.1 | - | 6 | 761 |
Gene Symbol: LOC100911293 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766401.1 | XP_008764623.1 | 756 |
Gene Symbol: LOC691658 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001079175.5 | XP_001079175.2 | 5 | 668 | ||
XM_003750532.3 | XP_003750580.1 | 6 | 790 | |||
XM_006243318.2 | XP_006243380.1 | 6 | 998 | |||
XM_006243319.2 | XP_006243381.1 | 5 | 879 |