Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tex9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729938.4 | XP_002729984.1 | 9 | 1707 | ||
XM_006243344.2 | XP_006243406.1 | 9 | 1674 | |||
GenBank mRNA | BC160875.1 | - | 8 | 1116 |