Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Leo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005548.1 | NP_001005548.1 | 2 | 146 | ||
GenBank mRNA | BC082098.1 | - | 2 | 146 | 7134452 | |
FQ229860.1 | - | 2 | 147 |