Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nup62cl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006234254.2 | XP_006234316.1 | 4 | 533 | ||
XM_006234255.2 | XP_006234317.1 | 4 | 427 | |||
XM_006234257.2 | XP_006234319.1 | 4 | 562 | |||
XM_008773468.1 | XP_008771690.1 | 4 | 481 | |||
XM_008773470.1 | XP_008771692.1 | 4 | 398 | |||
GenBank mRNA | BC160918.1 | - | 4 | 507 |