Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nup62cl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006234254.2 | XP_006234316.1 | 8, 9 | 988 | ||
XM_006234255.2 | XP_006234317.1 | 8, 9 | 882 | |||
XM_006234257.2 | XP_006234319.1 | 8, 9 | 1017 | |||
XM_008773468.1 | XP_008771690.1 | 8, 9 | 936 | |||
XM_008773470.1 | XP_008771692.1 | 8, 9 | 853 | |||
GenBank mRNA | BC160918.1 | - | 8, 9 | 962 |