Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100548.1 | NP_001094018.1 | 4, 5 | 626 | ||
XM_006243738.2 | XP_006243800.1 | 4, 5 | 880 | |||
XM_006243739.2 | XP_006243801.1 | 4, 5 | 518 | |||
GenBank mRNA | BC166477.1 | - | 4, 5 | 626 | 9102173 | |
FQ213299.1 | - | 4, 5 | 491 | |||
FQ226392.1 | - | 4, 5 | 508 | |||
FQ226478.1 | - | 4, 5 | 495 | |||
FQ226712.1 | - | 4, 5 | 492 | |||
FQ227608.1 | - | 4, 5 | 851 | |||
FQ230921.1 | - | 4, 5 | 512 | |||
FQ231080.1 | - | 4, 5 | 850 | |||
FQ232017.1 | - | 4, 5 | 493 | |||
FQ232808.1 | - | 4, 5 | 496 | |||
FQ235011.1 | - | 4, 5 | 493 |