Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100548.1 | NP_001094018.1 | 17 | 1670 | ||
XM_006243738.2 | XP_006243800.1 | 17 | 1924 | |||
XM_006243739.2 | XP_006243801.1 | 17 | 1562 | |||
GenBank mRNA | BC166477.1 | - | 17 | 1670 | 9102173 |