Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LUZP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142546.1 | NP_001136018.1 | 3 | 895 | ||
NM_033631.3 | NP_361013.3 | 4 | 961 | |||
XM_011542090.2 | XP_011540392.1 | 5 | 1445 | |||
XM_011542091.2 | XP_011540393.1 | 4 | 1379 | |||
XM_017002252.1 | XP_016857741.1 | 6 | 1619 | |||
GenBank mRNA | AK074153.1 | - | 1 | 556 | ||
AK093016.1 | - | 4 | 1379 | |||
BC002428.1 | - | 4 | 938 | |||
BC016175.1 | - | 4 | 948 | |||
BC051733.1 | - | 2 | 748 | 4838475 |