Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191819.1 | NP_001178748.1 | 8 | 1034 | ||
XM_006243963.2 | XP_006244025.1 | 8 | 1017 | |||
XM_008766630.1 | XP_008764852.1 | 9 | 1066 |