Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmtm8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198754.2 | NP_942049.1 | 2, 3 | 678 | ||
XM_006243964.2 | XP_006244026.1 | 2, 3 | 384 | |||
GenBank mRNA | AF538323.1 | - | 2, 3 | 481 | ||
BC088094.1 | - | 2, 3 | 678 | 7325894 |