Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EVI5 SIRNA Assays ›
Gene Symbol:
EVI5
Gene Name:
ecotropic viral integration site 5
Gene Aliases:
EVI-5, NB4S
Chromosome Location:
Chr.1: 92508696 - 92792404 on Build GRCh38
UniGene ID:
Hs.594434
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s15376
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EVI5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001308248.1
NP_001295177.1
18
2109
NM_005665.5
NP_005656.4
17
2076
XM_017002269.1
XP_016857758.1
20
2335
XM_017002270.1
XP_016857759.1
20
2646
XM_017002271.1
XP_016857760.1
19
2254
XM_017002272.1
XP_016857761.1
20
2335
XM_017002273.1
XP_016857762.1
20
2561
XM_017002274.1
XP_016857763.1
20
2310
XM_017002275.1
XP_016857764.1
20
2298
XM_017002276.1
XP_016857765.1
19
2478
XM_017002277.1
XP_016857766.1
18
2107
XM_017002278.1
XP_016857767.1
19
2188
XM_017002279.1
XP_016857768.1
19
2247
XM_017002280.1
XP_016857769.1
19
2478
XM_017002281.1
XP_016857770.1
18
2409
XM_017002282.1
XP_016857771.1
20
2335
XM_017002283.1
XP_016857772.1
19
2254
XM_017002284.1
XP_016857773.1
18
2330
XM_017002285.1
XP_016857774.1
19
2477
XM_017002286.1
XP_016857775.1
18
2248
XM_017002287.1
XP_016857776.1
19
2329
XM_017002288.1
XP_016857777.1
17
1871
GenBank mRNA
AB209513.1
-
13
1255
AB449875.1
-
18
2099
AF008915.1
-
17
2077
Back To Top
More Information
Panther Classification:
Molecular Function -
GTPase-activating protein
Gene Ontology Categories:
Function(s)
Process(es)
intracellular protein transport
cell cycle
multicellular organism development
cell proliferation
regulation of vesicle fusion
retrograde transport, endosome to Golgi
cell division
activation of GTPase activity
regulation of cilium assembly
GTPase activator activity
protein binding
Rab GTPase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-9xzrh:80/100.66.79.6:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline