Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EVI5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308248.1 | NP_001295177.1 | 7 | 1031 | ||
NM_005665.5 | NP_005656.4 | 7 | 1031 | |||
XM_017002269.1 | XP_016857758.1 | 8 | 1110 | |||
XM_017002270.1 | XP_016857759.1 | 8 | 1421 | |||
XM_017002271.1 | XP_016857760.1 | 7 | 1029 | |||
XM_017002272.1 | XP_016857761.1 | 8 | 1110 | |||
XM_017002273.1 | XP_016857762.1 | 8 | 1336 | |||
XM_017002274.1 | XP_016857763.1 | 8 | 1085 | |||
XM_017002275.1 | XP_016857764.1 | 8 | 1073 | |||
XM_017002276.1 | XP_016857765.1 | 7 | 1253 | |||
XM_017002277.1 | XP_016857766.1 | 7 | 1029 | |||
XM_017002278.1 | XP_016857767.1 | 8 | 1110 | |||
XM_017002279.1 | XP_016857768.1 | 7 | 1022 | |||
XM_017002280.1 | XP_016857769.1 | 7 | 1253 | |||
XM_017002281.1 | XP_016857770.1 | 7 | 1331 | |||
XM_017002282.1 | XP_016857771.1 | 8 | 1110 | |||
XM_017002283.1 | XP_016857772.1 | 7 | 1029 | |||
XM_017002284.1 | XP_016857773.1 | 7 | 1252 | |||
XM_017002285.1 | XP_016857774.1 | 7 | 1252 | |||
XM_017002286.1 | XP_016857775.1 | 6 | 1023 | |||
XM_017002287.1 | XP_016857776.1 | 7 | 1104 | |||
XM_017002288.1 | XP_016857777.1 | 5 | 646 | |||
GenBank mRNA | AB449875.1 | - | 7 | 1021 | ||
AF008915.1 | - | 7 | 1032 |