Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gltscr1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106888.1 | NP_001100358.1 | 3 | 142 | ||
XM_006244508.2 | XP_006244570.1 | 2 | 1880 | |||
XM_008766846.1 | XP_008765068.1 | 4 | 204 | |||
XM_008766847.1 | XP_008765069.1 | 3 | 168 |