Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106890.2 | NP_001100360.1 | 3 | 310 | ||
NM_001271131.1 | NP_001258060.1 | 3 | 266 | |||
XM_006244512.2 | XP_006244574.1 | 3 | 500 | |||
XM_008766849.1 | XP_008765071.1 | 4 | 433 | |||
XM_008766850.1 | XP_008765072.1 | 3 | 323 | |||
GenBank mRNA | FM100935.1 | - | 3 | 240 |