Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPHS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297575.1 | NP_001284504.1 | 4 | 573 | ||
NM_014625.3 | NP_055440.1 | 4 | 573 | |||
XM_005245483.3 | XP_005245540.1 | 2 | 418 | |||
XM_017002298.1 | XP_016857787.1 | 3 | 530 | |||
XM_017002299.1 | XP_016857788.1 | 4 | 608 | |||
GenBank mRNA | AJ279254.1 | - | 4 | 554 | ||
BC029141.1 | - | 4 | 573 | 5187188 |