Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prim2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024762.1 | NP_001019933.1 | 5 | 501 | ||
XM_006244686.2 | XP_006244748.1 | 3 | 330 | |||
XM_006244689.2 | XP_006244751.1 | 3 | 281 | |||
XM_006244690.2 | XP_006244752.1 | 3 | 347 | |||
XM_006244691.1 | XP_006244753.1 | 3 | 348 | |||
GenBank mRNA | AJ011607.1 | - | 2 | 185 | ||
BC097454.1 | - | 5 | 501 | 7456950 |