Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100549.1 | NP_001094019.1 | 5 | 507 | ||
XM_008767019.1 | XP_008765241.1 | 5 | 518 | |||
GenBank mRNA | BC088242.1 | - | 4 | 319 | ||
BC158704.1 | - | 5 | 475 | 9030006 | ||
CV106852.1 | - | 5 | 507 |