Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALMS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015120.4 | NP_055935.4 | 10 | 8685 | ||
GenBank mRNA | AB002326.3 | - | 3 | 2374 | ||
AJ417593.2 | - | 10 | 8685 | |||
AX751581.1 | - | 9 | 8559 | |||
BC035025.2 | - | 3 | 1373 | |||
BC050330.1 | - | 3 | 1373 | |||
BX647706.1 | - | 3 | 2598 |