Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOGS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146158.1 | NP_001139630.1 | 3 | 405 | ||
NM_006302.2 | NP_006293.2 | 2 | 731 | |||
GenBank mRNA | AK292553.1 | - | 2 | 677 | ||
AK307991.1 | - | 3 | 391 | |||
AK313355.1 | - | 2 | 731 | |||
BC028337.1 | - | 2 | 712 | 5110525 | ||
BC117250.1 | - | 2 | 604 | 40125801 | ||
BC117252.1 | - | 2 | 604 | 40125803 | ||
DC388420.1 | - | 3 | 377 | |||
X87237.1 | - | 2 | 700 |