Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOGS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146158.1 | NP_001139630.1 | 5 | 1391 | ||
NM_006302.2 | NP_006293.2 | 4 | 1717 | |||
XM_017004876.1 | XP_016860365.1 | 3 | 1480 | |||
XM_017004877.1 | XP_016860366.1 | 4 | 1155 | |||
GenBank mRNA | AK292553.1 | - | 4 | 1663 | ||
AK292719.1 | - | 2 | 1235 | |||
AK307991.1 | - | 5 | 1377 | |||
AK313355.1 | - | 4 | 1717 | |||
BC028337.1 | - | 4 | 1698 | 5110525 | ||
BC117250.1 | - | 4 | 1590 | 40125801 | ||
BC117252.1 | - | 4 | 1590 | 40125803 | ||
X87237.1 | - | 4 | 1686 |