Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Carf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106915.1 | NP_001100385.1 | 11, 12 | 1236 | ||
XM_006244969.1 | XP_006245031.1 | 8, 9 | 1098 | |||
XM_006244970.2 | XP_006245032.1 | 9, 10 | 1384 | |||
XM_006244971.1 | XP_006245033.1 | 7, 8 | 939 | |||
XM_006244973.2 | XP_006245035.1 | 6, 7 | 660 | |||
XM_006244974.2 | XP_006245036.1 | 7, 8 | 1624 |