Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fer | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106928.1 | NP_001100398.1 | 14 | 1782 | ||
XM_006245600.2 | XP_006245662.1 | 19 | 2514 | |||
XM_008767367.1 | XP_008765589.1 | 19 | 3290 | |||
XM_008767369.1 | XP_008765591.1 | 20 | 2652 | |||
XM_008767370.1 | XP_008765592.1 | 19 | 3550 | |||
XM_008767371.1 | XP_008765593.1 | 19 | 3287 | |||
XM_008767372.1 | XP_008765594.1 | 12 | 1427 | |||
GenBank mRNA | X13412.1 | - | 9 | 868 |