Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003469.4 | NP_003460.2 | 2 | 1728 | ||
GenBank mRNA | AK298828.1 | - | 3 | 1169 | ||
AK312452.1 | - | 2 | 1589 | |||
BC022509.1 | - | 2 | 1636 | 4820104 | ||
M25756.1 | - | 2 | 1561 |