Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppil2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017383.1 | NP_001017383.1 | 12 | 886 | ||
XM_008768892.1 | XP_008767114.1 | 12 | 922 | |||
GenBank mRNA | BC085890.1 | - | 12 | 886 | 7192457 |