Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC689479 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750610.3 | XP_003750658.2 | 1 | 495 | ||
XM_008766736.1 | XP_008764958.1 | 1 | 496 |
Gene Symbol: LOC302192 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006253275.2 | XP_006253337.1 | 151 | |||
XM_006253276.2 | XP_006253338.1 | 112 | ||||
GenBank mRNA | BC082094.1 | - | 90 | |||
BC100254.1 | - | 92 | 7456988 |
Gene Symbol: LOC681180 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006253262.2 | XP_006253324.1 | 1 | 151 | ||
XM_006253263.2 | XP_006253325.1 | 1 | 112 |