Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3bgrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173339.1 | NP_001166810.1 | 127 | |||
GenBank mRNA | BC169023.1 | - | 35 | 9025124 | ||
EV766621.1 | - | 1 | 127 |