Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPKAPK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243925.1 | NP_001230854.1 | 6 | 774 | ||
NM_001243926.1 | NP_001230855.1 | 8 | 1253 | |||
NM_004635.4 | NP_004626.1 | 6 | 759 | |||
GenBank mRNA | AB451303.1 | - | 5 | 611 | ||
AB451434.1 | - | 5 | 611 | |||
AK290969.1 | - | 6 | 730 | |||
AK313565.1 | - | 5 | 676 | |||
BC001662.2 | - | 6 | 720 | 2988489 | ||
BC007591.1 | - | 6 | 696 | 3345889 | ||
BC010407.2 | - | 6 | 718 | 3833667 | ||
BC068497.1 | - | 8 | 1207 | |||
U09578.1 | - | 6 | 702 | |||
U43784.1 | - | 5 | 633 |