Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdzd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106945.1 | NP_001100415.1 | 6, 7 | 582 | ||
XM_006257068.2 | XP_006257130.1 | 6, 7 | 488 | |||
XM_006257070.2 | XP_006257132.1 | 5, 6 | 423 | |||
GenBank mRNA | BC168740.1 | - | 5, 6 | 346 | 6918646 |