Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdzd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106945.1 | NP_001100415.1 | 5 | 487 | ||
XM_006257068.2 | XP_006257130.1 | 5 | 393 | |||
XM_006257070.2 | XP_006257132.1 | 4 | 328 | |||
GenBank mRNA | BC168740.1 | - | 4 | 251 | 6918646 |