Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGC114483 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024880.1 | NP_001020051.1 | 3 | 672 | ||
XM_006257008.2 | XP_006257070.1 | 4 | 1023 | |||
XM_006257009.2 | XP_006257071.1 | 4 | 743 | |||
XM_006257010.2 | XP_006257072.1 | 3 | 656 | |||
GenBank mRNA | BC097433.1 | - | 3 | 672 | 7455406 |