Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SLMAP SIRNA Assays ›
Gene Symbol:
SLMAP
Gene Name:
sarcolemma associated protein
Gene Aliases:
SLAP
Chromosome Location:
Chr.3: 57756221 - 57930013 on Build GRCh38
UniGene ID:
Hs.476432
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s15435
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SLMAP
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001304420.2
NP_001291349.1
19
2401
NM_001304421.2
NP_001291350.1
17
2287
NM_001304422.2
NP_001291351.1
7
883
NM_001304423.2
NP_001291352.1
5
685
NM_001311178.1
NP_001298107.1
7
883
NM_001311179.1
NP_001298108.1
6
760
NM_007159.4
NP_009090.2
18
2350
XM_005265456.3
XP_005265513.1
21
3288
XM_005265457.3
XP_005265514.1
21
3282
XM_005265458.3
XP_005265515.1
21
3288
XM_005265459.3
XP_005265516.1
20
3237
XM_005265460.3
XP_005265517.1
20
3237
XM_005265461.3
XP_005265518.1
20
3231
XM_005265462.3
XP_005265519.1
20
3225
XM_005265464.3
XP_005265521.1
20
3219
XM_005265465.3
XP_005265522.1
19
3186
XM_005265466.3
XP_005265523.1
19
3180
XM_005265467.3
XP_005265524.1
19
3174
XM_005265468.3
XP_005265525.1
20
3165
XM_005265470.3
XP_005265527.1
18
3123
XM_005265471.3
XP_005265528.1
18
3117
XM_005265472.3
XP_005265529.1
18
3051
XM_006713326.2
XP_006713389.1
18
3057
XM_011534103.1
XP_011532405.1
19
3174
XM_011534104.1
XP_011532406.1
18
3063
XM_011534105.1
XP_011532407.1
17
3000
XM_011534106.1
XP_011532408.1
7
758
XM_017007146.1
XP_016862635.1
21
3282
XM_017007147.1
XP_016862636.1
20
3231
XM_017007148.1
XP_016862637.1
20
3237
XM_017007149.1
XP_016862638.1
20
3231
XM_017007150.1
XP_016862639.1
20
3231
XM_017007151.1
XP_016862640.1
19
3186
XM_017007152.1
XP_016862641.1
19
3180
XM_017007153.1
XP_016862642.1
19
3168
XM_017007154.1
XP_016862643.1
19
3168
XM_017007155.1
XP_016862644.1
20
3159
XM_017007156.1
XP_016862645.1
20
3165
XM_017007157.1
XP_016862646.1
20
3159
XM_017007158.1
XP_016862647.1
18
3123
XM_017007159.1
XP_016862648.1
19
3114
XM_017007160.1
XP_016862649.1
18
3117
XM_017007161.1
XP_016862650.1
19
3108
XM_017007162.1
XP_016862651.1
19
3114
XM_017007163.1
XP_016862652.1
19
3108
XM_017007164.1
XP_016862653.1
18
3057
XM_017007165.1
XP_016862654.1
18
3045
XM_017007166.1
XP_016862655.1
18
3045
XM_017007167.1
XP_016862656.1
17
3000
XM_017007168.1
XP_016862657.1
17
2994
XM_017007170.1
XP_016862659.1
7
758
GenBank mRNA
AB046821.1
-
16
1591
AF100750.1
-
7
827
AF304450.1
-
7
756
AK022561.1
-
2
294
AK300106.1
-
5
608
AK302934.1
-
6
635
AK304493.1
-
7
810
AK316436.1
-
7
751
AL834538.1
-
3
424
BC114627.1
-
17
1985
40081892
BC115701.1
-
19
2099
40081893
CR627321.1
-
6
617
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
muscle contraction
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-slhn9:80/100.66.78.111:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline