Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MANF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006010.5 | NP_006001.4 | 2 | 230 | ||
GenBank mRNA | AK292243.1 | - | 2 | 143 | ||
BC007282.1 | - | 2 | 126 | |||
BC113588.1 | - | 2 | 153 | 8322640 | ||
BC113590.1 | - | 2 | 153 | 8322642 | ||
BE514267.1 | - | 2 | 238 | |||
BT007110.1 | - | 2 | 124 | |||
CN410909.1 | - | 2 | 172 | |||
CN410911.1 | - | 2 | 219 | |||
M83751.1 | - | 2 | 412 |