Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135562.1 | NP_001129034.1 | 4 | 354 | ||
NM_021262.2 | NP_067085.1 | 4 | 321 | |||
XM_006239965.2 | XP_006240027.1 | 4 | 350 | |||
XM_008764572.1 | XP_008762794.1 | 4 | 354 | |||
GenBank mRNA | AF171072.1 | - | 4 | 284 | ||
BC062229.1 | - | 4 | 321 | 6918194 | ||
BC158763.1 | - | 4 | 354 | 7109165 | ||
BC161990.1 | - | 4 | 300 | 9032684 | ||
FQ213063.1 | - | 4 | 301 | |||
FQ214806.1 | - | 3 | 189 | |||
FQ215147.1 | - | 4 | 292 |