Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286457.1 | NP_001273386.1 | 16 | 1815 | ||
NM_001286458.1 | NP_001273387.1 | 17 | 1990 | |||
NM_001321858.1 | NP_001308787.1 | 16 | 1741 | |||
NM_003470.2 | NP_003461.2 | 16 | 1946 | |||
NR_135826.1 | 16 | 1728 | ||||
XM_017023652.1 | XP_016879141.1 | 16 | 2374 | |||
XM_017023653.1 | XP_016879142.1 | 16 | 1810 | |||
GenBank mRNA | AK302481.1 | - | 17 | 1916 | ||
AK302771.1 | - | 16 | 1747 | |||
AK302872.1 | - | 16 | 1728 | |||
AK302912.1 | - | 17 | 1934 | |||
AK316441.1 | - | 16 | 1816 | |||
AY376241.1 | - | 16 | 1777 | |||
Z72499.1 | - | 16 | 1946 |