Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004637.5 | NP_004628.4 | 3 | 365 | ||
GenBank mRNA | AF498942.1 | - | 2 | 133 | ||
AK094449.1 | - | 3 | 318 | |||
AK225177.1 | - | 2 | 119 | |||
AK225691.1 | - | 2 | 112 | |||
AK290721.1 | - | 3 | 314 | |||
AK298344.1 | - | 3 | 288 | |||
AK308934.1 | - | 3 | 308 | |||
AU122058.1 | - | 3 | 365 | |||
BC008721.2 | - | 3 | 308 | 2821435 | ||
BC013728.1 | - | 2 | 148 | 3882701 | ||
BX537775.1 | - | 1 | 81 | |||
U44104.1 | - | 2 | 133 |